missing translation for 'onlineSavingsMsg'
Learn More

3-Trimethylsilylpropynal, 97%

CAS: 2975-46-4 | C6H10OSi | 126.23 g/mol

$174.93 - $723.78

Chemical Identifiers

CAS 2975-46-4
Molecular Formula C6H10OSi
Molecular Weight (g/mol) 126.23
MDL Number MFCD01103709
InChI Key LJRWLSKYGWLYIM-UHFFFAOYSA-N
Synonym 3-trimethylsilylpropynal, 3-trimethylsilyl-2-propynal, 3-trimethylsilyl propiolaldehyde, 3-trimethylsilyl propynal, trimethylsilyl propiolaldehyde, 2-propynal, 3-trimethylsilyl, trimethylsilylpropynal, 3-trimethylsilyl prop-2-ynal
PubChem CID 2760865
IUPAC Name 3-trimethylsilylprop-2-ynal
SMILES C[Si](C)(C)C#CC=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA4476303
View Documents
Thermo Scientific Chemicals
04476303
1 g
Each for $174.93
Only null left
 
AA4476306
View Documents
Thermo Scientific Chemicals
04476306
5 g
Each for $723.78
Only null left
 
Description

Description

3-Trimethylsilylpropynal acts as an important organic intermediate. It is used in agrochemicals, pharmaceuticals and dyestuff field. Further, it is used in the preparation of [3-(trimethylsilyl)-2-propinyliden]-malononitrile.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon
Specifications

Chemical Identifiers

2975-46-4
126.23
LJRWLSKYGWLYIM-UHFFFAOYSA-N
2760865
C[Si](C)(C)C#CC=O
C6H10OSi
MFCD01103709
3-trimethylsilylpropynal, 3-trimethylsilyl-2-propynal, 3-trimethylsilyl propiolaldehyde, 3-trimethylsilyl propynal, trimethylsilyl propiolaldehyde, 2-propynal, 3-trimethylsilyl, trimethylsilylpropynal, 3-trimethylsilyl prop-2-ynal
3-trimethylsilylprop-2-ynal

Specifications

2975-46-4
0.86 g/mL
29°C (84°F)
1.4435
MFCD01103709
UN1993
3-trimethylsilylpropynal, 3-trimethylsilyl-2-propynal, 3-trimethylsilyl propiolaldehyde, 3-trimethylsilyl propynal, trimethylsilyl propiolaldehyde, 2-propynal, 3-trimethylsilyl, trimethylsilylpropynal, 3-trimethylsilyl prop-2-ynal
C[Si](C)(C)C#CC=O
126.23
126.23
Liquid
Colorless to Yellow
58°C to 60°C (37.5 mmHg)
C6H10OSi
(CH3)3SiC≡CCHO
1 g
Air and moisture sensitive
LJRWLSKYGWLYIM-UHFFFAOYSA-N
3-trimethylsilylprop-2-ynal
2760865
97%
3-Trimethylsilylpropynal
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c

H226-H315-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

TSCA : No

storageNote1 :

  • Packaged under Argon

Recommended Storage : Keep cold; Store under Nitrogen

SDS
Documents

Documents

RUO – Research Use Only