missing translation for 'onlineSavingsMsg'
Learn More

3-(Trimethylsilylethynyl)thiophene 98.0+%, TCI America™

Catalog No. T31901G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T31901G 1 g
T31905G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T31901G Supplier TCI America Supplier No. T31901G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 130995-13-0
Molecular Formula C9H12SSi
Molecular Weight (g/mol) 180.34
MDL Number MFCD04039959
InChI Key XJUQFUWWPCIZRB-UHFFFAOYSA-N
Synonym trimethyl thiophen-3-ylethynyl silane, 3-trimethylsilylethynyl thiophene, 3-trimethylsilyl ethynyl thiophene, trimethyl 2-thiophen-3-ylethynyl silane, 3-thienylethynyl trimethylsilane, 3-trimethylsilylethynyl thiophene 97, trimethyl 2-thiophen-3-yl ethynyl silane, 3-trimethylsilyl-ethynyl thiophene, trimethyl-2-3-thienyl ethynyl silane, trimethyl thiophen-3-yl ethynyl silane
PubChem CID 3654101
IUPAC Name trimethyl(2-thiophen-3-ylethynyl)silane
SMILES C[Si](C)(C)C#CC1=CSC=C1

Specifications

Color Yellow
Quantity 1 g
Formula Weight 180.34
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-(Trimethylsilylethynyl)thiophene