missing translation for 'onlineSavingsMsg'
Learn More

3-(Trimethylsilyl)propargyl bromide, 97%

Catalog No. AC399840050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC399840050 5 g
AC399840010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC399840050 Supplier Thermo Scientific Chemicals Supplier No. 399840050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 38002-45-8 | C6H11BrSi | 191.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 38002-45-8
Molecular Formula C6H11BrSi
Molecular Weight (g/mol) 191.14
MDL Number MFCD00134460
InChI Key GAPRPFRDVCCCHR-UHFFFAOYSA-N
Synonym 3-bromo-1-trimethylsilyl-1-propyne, 3-bromoprop-1-yn-1-yl trimethylsilane, 3-bromo-1-propynyl trimethylsilane, 3-trimethylsilyl propargyl bromide, 3-bromoprop-1-ynyl trimethyl silane, silane, 3-bromo-1-propynyl trimethyl, 3-bromoprop-1-yn-1-yl trimethyl silane, acmc-1cthe, ksc496i3r
PubChem CID 642589
IUPAC Name 3-bromoprop-1-ynyl(trimethyl)silane
SMILES C[Si](C)(C)C#CCBr

Specifications

Density 1.3510g/mL
Boiling Point 44°C to 45°C (2.0 mmHg)
Flash Point 62°C
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Linear Formula (CH3)3SiC∽CCH2Br
Quantity 5 g
Specific Gravity 1.351
Solubility Information Solubility in water: insoluble
Formula Weight 191.14
Percent Purity 97%
Chemical Name or Material 3-(Trimethylsilyl)propargyl bromide
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Avoid breathing dust/fume/

RUO – Research Use Only