missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(Trimethylsilyl)propargyl bromide, 97%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Density | 1.3510g/mL |
| Boiling Point | 44°C to 45°C (2.0 mmHg) |
| Flash Point | 62°C |
| Assay Percent Range | 96% min. (GC) |
| Packaging | Glass Bottle |
| Linear Formula | (CH3)3SiC∽CCH2Br |
| Quantity | 1 g |
| Specific Gravity | 1.351 |
| Solubility Information | Solubility in water: insoluble |
| Formula Weight | 191.14 |
| Show More |
Safety and Handling
| missing translation for 'healthHazard1' | GHS Signal Word: Warning |
| missing translation for 'healthHazard2' | GHS H Statement May cause respiratory irritation. Causes skin irritation. Causes serious eye irritation. |
| missing translation for 'healthHazard3' | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. If skin irritation occurs: Get medical advice/attention. If eye irritation persists: Get medical advice/attention. Avoid breathing dust/fume/ |
RUO – Research Use Only