missing translation for 'onlineSavingsMsg'
Learn More

3-(Trimethylsilyl)propargyl alcohol, 98%

Catalog No. p-7045161
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0964903 1 g
AAL0964906 5 g
AAL0964914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL0964903 Supplier Thermo Scientific Chemicals Supplier No. L0964903

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 5272-36-6 | C6H12OSi | 128.25 g/mol

3-(Trimethylsilyl)propargyl alcohol is used as a reagent to synthesize Isbogrel, a Thromboxane A 2 (TXA 2) synthase inhibitor and also a TXA 2 receptor antagonist that has potential to treat asthma.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-(Trimethylsilyl)propargyl alcohol is used as a reagent to synthesize Isbogrel, a Thromboxane A 2 (TXA 2) synthase inhibitor and also a TXA 2 receptor antagonist that has potential to treat asthma.

Solubility
Not miscible with water.

Notes
Moisture Sensitive. Store away from moisture and oxidizing agents. Protect from heat. Store under dry inert gas. Protect from humidity and water. Incompatible with oxidizing agents, heat and moisture.

Chemical Identifiers

CAS 5272-36-6
Molecular Formula C6H12OSi
Molecular Weight (g/mol) 128.25
MDL Number MFCD00002913
InChI Key ZVGCJDPEKKEYES-UHFFFAOYSA-N
Synonym 3-trimethylsilyl-2-propyn-1-ol, 3-trimethylsilyl propargyl alcohol, 2-propyn-1-ol, 3-trimethylsilyl, 3-trimethylsilyl prop-2-yn-1-ol, 1-trimethylsilylpropargyl alcohol, acmc-209l1b, 3-trimethylsilylpropargyl alcohol, 3-trimethylsilanyl-prop-2-yn-1-ol, 3-trimethylsilyl-2-propyn-1-ol #
PubChem CID 78930
IUPAC Name 3-trimethylsilylprop-2-yn-1-ol
SMILES C[Si](C)(C)C#CCO

Specifications

Boiling Point 170°C to 172°C
Quantity 1 g
Linear Formula (CH3)3SiC≡CCH2OH
UN Number UN2810
Beilstein 1902505
Flash Point 66°C (150°F)
Solubility Information Not miscible with water.
Formula Weight 128.25
Percent Purity ≥98%
Refractive Index 1.451
Sensitivity Moisture sensitive
Density 0.865
Chemical Name or Material 3-(Trimethylsilyl)propargyl alcohol
Show More Show Less
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H301-H227-H315-H319-H335
Toxic if swallowed.
Combustible liquid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber 226-094-5
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only