accessibility menu, dialog, popup

UserName

CAS RN 5272-36-6

IUPAC Name: 3-(trimethylsilyl)prop-2-yn-1-ol
Synonyms: 3-(trimethylsilyl)prop-2-yn-1-ol
Molecular Weight (g/mol): 128.25
Molecular Formula: C6H12OSi
InChi Key: ZVGCJDPEKKEYES-UHFFFAOYSA-N
SMILES: C[Si](C)(C)C#CCO
Percent Purity 
  • (1)
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (4)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)