missing translation for 'onlineSavingsMsg'
Learn More

3-[(Trimethylsilyl)ethynyl]benzonitrile, 97%

Catalog No. p-7036681
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5501203 1 g
AAH5501206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5501203 Supplier Thermo Scientific Chemicals Supplier No. H5501203

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 190771-22-3 | C12H13NSi | 199.328 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 190771-22-3
Molecular Formula C12H13NSi
Molecular Weight (g/mol) 199.328
MDL Number MFCD09260494
InChI Key QEZDMZVLNWSJSA-UHFFFAOYSA-N
Synonym 3-trimethylsilyl ethynyl benzonitrile, 3-2-trimethylsilyl ethynyl benzonitrile, benzonitrile,3-2-trimethylsilyl ethynyl, benzonitrile, 3-2-trimethylsilyl ethynyl, 3-trimethylsilanylethynylbenzonitrile, 3-trimethylsilanylethynyl-benzonitrile
PubChem CID 22570318
IUPAC Name 3-(2-trimethylsilylethynyl)benzonitrile
SMILES C[Si](C)(C)C#CC1=CC(=CC=C1)C#N

Specifications

Melting Point 42°C to 47°C
Quantity 1 g
Formula Weight 199.33
Percent Purity 97%
Chemical Name or Material 3-[(Trimethylsilyl)ethynyl]benzonitrile
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only