CAS RN 190771-22-3
IUPAC Name:
3-[2-(trimethylsilyl)ethynyl]benzonitrile
Synonyms:
3-[2-(trimethylsilyl)ethynyl]benzonitrile
Molecular Weight (g/mol):
199.33
Molecular Formula:
C12H13NSi
InChi Key:
QEZDMZVLNWSJSA-UHFFFAOYSA-N
SMILES:
C[Si](C)(C)C#CC1=CC(=CC=C1)C#N
1
–
1
of
1
results
1
Promotions Available
3-[(Trimethylsilyl)ethynyl]benzonitrile, 97%
3-[(Trimethylsilyl)ethynyl]benzonitrile, 97%
CAS: 190771-22-3 Molecular Formula: C12H13NSi Molecular Weight (g/mol): 199.328 MDL Number: MFCD09260494 InChI Key: QEZDMZVLNWSJSA-UHFFFAOYSA-N Synonym: 3-trimethylsilyl ethynyl benzonitrile,3-2-trimethylsilyl ethynyl benzonitrile,benzonitrile,3-2-trimethylsilyl ethynyl,benzonitrile, 3-2-trimethylsilyl ethynyl,3-trimethylsilanylethynylbenzonitrile,3-trimethylsilanylethynyl-benzonitrile PubChem CID: 22570318 IUPAC Name: 3-(2-trimethylsilylethynyl)benzonitrile SMILES: C[Si](C)(C)C#CC1=CC(=CC=C1)C#N