missing translation for 'onlineSavingsMsg'
Learn More

(3-Trimethylsilyl-2-propynyl)triphenylphosphonium Bromide 98.0+%, TCI America™

Catalog No. T14985G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T14985G 5 g
T14981G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T14985G Supplier TCI America Supplier No. T14985G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 42134-49-6
Molecular Formula C24H26BrPSi
Molecular Weight (g/mol) 453.435
MDL Number MFCD00012029
InChI Key PBSHVEOONSKWJF-UHFFFAOYSA-M
Synonym 3-trimethylsilyl-2-propynyl triphenylphosphonium bromide, triphenyl 3-trimethylsilyl prop-2-yn-1-yl phosphonium bromide, triphenyl 3-trimethylsilyl prop-2-yn-1-yl phosphanium bromide, phosphonium, triphenyl 3-trimethylsilyl-2-propyn-1-yl-, bromide 1:1, acmc-209jnp, trimethylsilylpropargyl triphenylphosphonium bromide, 3-trimethylsilyl-2-propynyl triphenylphosphoniumbromide, triphenyl 3-trimethylsilylprop-2-ynyl phosphanium bromide, 3-trimethylsilyl-2-propyn-1-yltriphenylphosphonium bromide
PubChem CID 2723674
IUPAC Name triphenyl(3-trimethylsilylprop-2-ynyl)phosphanium;bromide
SMILES C[Si](C)(C)C#CC[P+](C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3.[Br-]

Specifications

Quantity 5 g
Formula Weight 453.43
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material (3-Trimethylsilyl-2-propynyl)triphenylphosphonium Bromide