missing translation for 'onlineSavingsMsg'
Learn More

3-Trimethylsilyl-2-oxazolidinone 98.0+%, TCI America™

Catalog No. T15355G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T15355G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T15355G Supplier TCI America Supplier No. T15355G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 43112-38-5
Molecular Formula C6H13NO2Si
Molecular Weight (g/mol) 159.26
MDL Number MFCD00010197
InChI Key AUKCYOUETBBMFV-UHFFFAOYSA-N
Synonym 3-trimethylsilyl-2-oxazolidinone, 3-trimethylsilyl oxazolidin-2-one, n-trimethylsilyl-2-oxazolidinone, 3-trimethylsilyl-2-oxazolidone, acmc-20amyd, pubchem24175, 2-oxazolidinone,3-trimethylsilyl, trimethyl 2-oxo-3-oxazolidinyl silane, 2-oxo-3-trimethylsilyl-1,3-oxazolidine
PubChem CID 123514
IUPAC Name 3-trimethylsilyl-1,3-oxazolidin-2-one
SMILES C[Si](C)(C)N1CCOC1=O

Specifications

Color Yellow
Boiling Point 122°C
Quantity 5 g
Formula Weight 159.26
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Trimethylsilyl-2-oxazolidinone [Trimethylsilylating Reagent]
TSCA No
Recommended Storage Freezer