CAS RN 43112-38-5
IUPAC Name:
3-(trimethylsilyl)-1,3-oxazolidin-2-one
Synonyms:
3-(trimethylsilyl)-1,3-oxazolidin-2-one
Molecular Weight (g/mol):
159.26
Molecular Formula:
C6H13NO2Si
InChi Key:
AUKCYOUETBBMFV-UHFFFAOYSA-N
SMILES:
C[Si](C)(C)N1CCOC1=O
1
–
1
of
1
results
1
3-Trimethylsilyl-2-oxazolidinone 98.0+%, TCI America™
CAS: 43112-38-5 Molecular Formula: C6H13NO2Si Molecular Weight (g/mol): 159.26 MDL Number: MFCD00010197 InChI Key: AUKCYOUETBBMFV-UHFFFAOYSA-N Synonym: 3-trimethylsilyl-2-oxazolidinone,3-trimethylsilyl oxazolidin-2-one,n-trimethylsilyl-2-oxazolidinone,3-trimethylsilyl-2-oxazolidone,acmc-20amyd,pubchem24175,2-oxazolidinone,3-trimethylsilyl,trimethyl 2-oxo-3-oxazolidinyl silane,2-oxo-3-trimethylsilyl-1,3-oxazolidine PubChem CID: 123514 IUPAC Name: 3-trimethylsilyl-1,3-oxazolidin-2-one SMILES: C[Si](C)(C)N1CCOC1=O