missing translation for 'onlineSavingsMsg'
Learn More

3-(Trimethylsilyl)-2-naphthyl Trifluoromethanesulfonate 95.0+%, TCI America™

Catalog No. T24661G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T24661G 1 g
T24665G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T24661G Supplier TCI America Supplier No. T24661G
Only null left

Chemical Identifiers

CAS 780820-43-1
Molecular Formula C14H15F3O3SSi
Molecular Weight (g/mol) 348.411
InChI Key OSZYFPWWHQEAGW-UHFFFAOYSA-N
Synonym 3-(Trimethylsilyl)-2-naphthyl Triflate, Trifluoromethanesulfonic Acid 3-(Trimethylsilyl)-2-naphthyl Ester
PubChem CID 44629848
IUPAC Name (3-trimethylsilylnaphthalen-2-yl) trifluoromethanesulfonate
SMILES C[Si](C)(C)C1=CC2=CC=CC=C2C=C1OS(=O)(=O)C(F)(F)F

Specifications

Color Yellow
Quantity 1 g
UN Number 3265
Formula Weight 348.41
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-(Trimethylsilyl)-2-naphthyl Trifluoromethanesulfonate
TSCA No
Recommended Storage Refrigerator