missing translation for 'onlineSavingsMsg'
Learn More

3-(Trimethoxysilyl)propyl Methacrylate (stabilized with BHT) 98.0+%, TCI America™

Supplier:  TCI America M0725100ML

 View more versions of this product

Catalog No. M0725100ML

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

3-(Trimethoxysilyl)propyl Methacrylate (stabilized with BHT)
Colorless
C10H20O5Si
100 mL
XDLMVUHYZWKMMD-UHFFFAOYSA-N
3-trimethoxysilylpropyl 2-methylprop-2-enoate
17318
≥98.0% (GC)
2530-85-0
110°C
MFCD00008593
3-trimethoxysilyl propyl methacrylate, 3-methacryloxypropyltrimethoxysilane, methacryloxypropyltrimethoxysilane, dynasylan memo, silicone a-174, union carbide a-174, mops-m, silane a-174, nuca 174, 2-propenoic acid, 2-methyl-, 3-trimethoxysilyl propyl ester
CC(=C)C(=O)OCCC[Si](OC)(OC)OC
248.35
248.35
Liquid

Chemical Identifiers

2530-85-0
248.35
XDLMVUHYZWKMMD-UHFFFAOYSA-N
17318
CC(=C)C(=O)OCCC[Si](OC)(OC)OC
C10H20O5Si
MFCD00008593
3-trimethoxysilyl propyl methacrylate, 3-methacryloxypropyltrimethoxysilane, methacryloxypropyltrimethoxysilane, dynasylan memo, silicone a-174, union carbide a-174, mops-m, silane a-174, nuca 174, 2-propenoic acid, 2-methyl-, 3-trimethoxysilyl propyl ester
3-trimethoxysilylpropyl 2-methylprop-2-enoate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-2076

RTECSNumber : UC0230000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions