missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethylthio)benzoic acid, 97%

Catalog No. AAA1997503
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1997503 1 g
AAA1997506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1997503 Supplier Thermo Scientific Chemicals Supplier No. A1997503
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 946-65-6 | C8H4F3O2S | 221.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 946-65-6
Molecular Formula C8H4F3O2S
Molecular Weight (g/mol) 221.17
MDL Number MFCD00236337
InChI Key IVGAPIVNQABETQ-UHFFFAOYSA-M
Synonym 3-trifluoromethylthio benzoic acid, 3-trifluoromethyl thio benzoic acid, 3-trifluoromethyl sulfanyl benzoic acid, 3-trifluoromethylsulfanyl benzoic acid, 3-trifluoromethylsulfanyl-benzoic acid, benzoic acid, 3-trifluoromethyl thio, 3-trifluoromethyl sulphanyl benzoic acid, 3-trifluoromethylthio benzoicacid, 3-carboxyphenyl trifluoromethyl sulphide
PubChem CID 688330
IUPAC Name 3-(trifluoromethylsulfanyl)benzoic acid
SMILES [O-]C(=O)C1=CC=CC(SC(F)(F)F)=C1

Specifications

Melting Point 71°C to 74°C
Quantity 1 g
Formula Weight 222.18
Percent Purity 97%
Chemical Name or Material 3-(Trifluoromethylthio)benzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only