missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethyl)thiobenzamide, 97%

CAS: 53515-17-6 | C8H6F3NS | 205.20 g/mol

Supplier:  Thermo Scientific Chemicals H2631106

 View more versions of this product

Catalog No. AAH2631106


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents and bases.
Specifications

Specifications

3-(Trifluoromethyl)thiobenzamide
C8H6F3NS
5 g
3-trifluoromethyl thiobenzamide, 3-trifluoromethylthiobenzamide, 3-trifluoromethyl benzenecarbothioamide, 3-trifluoromethyl benzene-1-carbothioamide, 3-trifluoromethyl-thiobenzamide, amino 3-trifluoromethyl phenyl methane-1-thione, acmc-20amye, pubchem5483, maybridge1_008312
WOJQBHSZMFRQIR-UHFFFAOYSA-N
3-(trifluoromethyl)benzenecarbothioamide
2777849
97%
53515-17-6
MFCD01567258
UN2811
Insoluble in water.
NC(=S)C1=CC(=CC=C1)C(F)(F)F
205.20
205.2

Chemical Identifiers

53515-17-6
205.20
WOJQBHSZMFRQIR-UHFFFAOYSA-N
2777849
NC(=S)C1=CC(=CC=C1)C(F)(F)F
C8H6F3NS
MFCD01567258
3-trifluoromethyl thiobenzamide, 3-trifluoromethylthiobenzamide, 3-trifluoromethyl benzenecarbothioamide, 3-trifluoromethyl benzene-1-carbothioamide, 3-trifluoromethyl-thiobenzamide, amino 3-trifluoromethyl phenyl methane-1-thione, acmc-20amye, pubchem5483, maybridge1_008312
3-(trifluoromethyl)benzenecarbothioamide
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only