missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethyl)thiobenzamide, 97%

Catalog No. AAH26311MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH26311MD 250 mg
AAH2631103 1 g
AAH2631106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH26311MD Supplier Thermo Scientific Chemicals Supplier No. H26311MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 53515-17-6 | C8H6F3NS | 205.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents and bases.

Chemical Identifiers

CAS 53515-17-6
Molecular Formula C8H6F3NS
Molecular Weight (g/mol) 205.20
MDL Number MFCD01567258
InChI Key WOJQBHSZMFRQIR-UHFFFAOYSA-N
Synonym 3-trifluoromethyl thiobenzamide, 3-trifluoromethylthiobenzamide, 3-trifluoromethyl benzenecarbothioamide, 3-trifluoromethyl benzene-1-carbothioamide, 3-trifluoromethyl-thiobenzamide, amino 3-trifluoromethyl phenyl methane-1-thione, acmc-20amye, pubchem5483, maybridge1_008312
PubChem CID 2777849
IUPAC Name 3-(trifluoromethyl)benzenecarbothioamide
SMILES NC(=S)C1=CC(=CC=C1)C(F)(F)F

Specifications

Quantity 250 mg
UN Number UN2811
Solubility Information Insoluble in water.
Formula Weight 205.2
Percent Purity 97%
Chemical Name or Material 3-(Trifluoromethyl)thiobenzamide
Hazard Category H302+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only