missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(Trifluoromethyl)styrene (stabilized with HQ) 97.0+%, TCI America™
$238.60 - $734.44
Chemical Identifiers
| CAS | 402-24-4 |
|---|---|
| Molecular Formula | C9H7F3 |
| Molecular Weight (g/mol) | 172.15 |
| MDL Number | MFCD00075481 |
| InChI Key | ARHOUOIHKWELMD-UHFFFAOYSA-N |
| Synonym | 3-trifluoromethyl styrene, 1-trifluoromethyl-3-vinylbenzene, 1-ethenyl-3-trifluoromethyl benzene, 3-cf3c6h4ch=ch2, 3-trifluoromethylstyren, 3-vinylbenzotrifluoride, m-trifluoromethylstyrene, pubchem15477, 3-trifluoromethylstyrene, 1-trifluoromethyl-3-vinyl-benzene |
| PubChem CID | 520986 |
| IUPAC Name | 1-ethenyl-3-(trifluoromethyl)benzene |
| SMILES | FC(F)(F)C1=CC=CC(C=C)=C1 |
Chemical Identifiers
| 402-24-4 | |
| 172.15 | |
| ARHOUOIHKWELMD-UHFFFAOYSA-N | |
| 520986 | |
| FC(F)(F)C1=CC=CC(C=C)=C1 |
| C9H7F3 | |
| MFCD00075481 | |
| 3-trifluoromethyl styrene, 1-trifluoromethyl-3-vinylbenzene, 1-ethenyl-3-trifluoromethyl benzene, 3-cf3c6h4ch=ch2, 3-trifluoromethylstyren, 3-vinylbenzotrifluoride, m-trifluoromethylstyrene, pubchem15477, 3-trifluoromethylstyrene, 1-trifluoromethyl-3-vinyl-benzene | |
| 1-ethenyl-3-(trifluoromethyl)benzene |
Specifications
| 402-24-4 | |
| 65°C | |
| MFCD00075481 | |
| 1993 | |
| ARHOUOIHKWELMD-UHFFFAOYSA-N | |
| 1-ethenyl-3-(trifluoromethyl)benzene | |
| 520986 | |
| ≥97.0% (GC) | |
| 3-(Trifluoromethyl)styrene (stabilized with HQ) |
| Yellow | |
| C9H7F3 | |
| 1 g | |
| 3-trifluoromethyl styrene, 1-trifluoromethyl-3-vinylbenzene, 1-ethenyl-3-trifluoromethyl benzene, 3-cf3c6h4ch=ch2, 3-trifluoromethylstyren, 3-vinylbenzotrifluoride, m-trifluoromethylstyrene, pubchem15477, 3-trifluoromethylstyrene, 1-trifluoromethyl-3-vinyl-benzene | |
| FC(F)(F)C1=CC=CC(C=C)=C1 | |
| 172.15 | |
| 172.15 | |
| Liquid |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator