missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethyl)styrene (stabilized with HQ) 97.0+%, TCI America™

Catalog No. T33541G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T33541G 1 g
T33545G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T33541G Supplier TCI America Supplier No. T33541G

Chemical Identifiers

CAS 402-24-4
Molecular Formula C9H7F3
Molecular Weight (g/mol) 172.15
MDL Number MFCD00075481
InChI Key ARHOUOIHKWELMD-UHFFFAOYSA-N
Synonym 3-trifluoromethyl styrene, 1-trifluoromethyl-3-vinylbenzene, 1-ethenyl-3-trifluoromethyl benzene, 3-cf3c6h4ch=ch2, 3-trifluoromethylstyren, 3-vinylbenzotrifluoride, m-trifluoromethylstyrene, pubchem15477, 3-trifluoromethylstyrene, 1-trifluoromethyl-3-vinyl-benzene
PubChem CID 520986
IUPAC Name 1-ethenyl-3-(trifluoromethyl)benzene
SMILES FC(F)(F)C1=CC=CC(C=C)=C1

Specifications

Color Yellow
Boiling Point 65°C
Quantity 1 g
UN Number 1993
Formula Weight 172.15
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-(Trifluoromethyl)styrene (stabilized with HQ)
TSCA No
Recommended Storage Refrigerator