missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethyl)pyridine 98.0+%, TCI America™

Supplier:  TCI America T278625G

 View more versions of this product

Catalog No. T278625G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

3-(Trifluoromethyl)pyridine
Yellow
C6H4F3N
25 g
3-trifluoromethyl pyridine, pyridine, 3-trifluoromethyl, 3-trifluoromethyl-pyridine, m-trifluoromethylpyridine, alpha,alpha,alpha-trifluoro-3-picoline, pubchem8025, 3-cf3-pyridine, 5-trifluoromethylpyridine
C1=CC(=CN=C1)C(F)(F)F
147.1
147.10
Liquid
3796-23-4
113°C
MFCD00100032
1993
JTZSFNHHVULOGJ-UHFFFAOYSA-N
3-(trifluoromethyl)pyridine
77417
≥98.0% (GC)

Chemical Identifiers

3796-23-4
147.1
JTZSFNHHVULOGJ-UHFFFAOYSA-N
77417
C1=CC(=CN=C1)C(F)(F)F
C6H4F3N
MFCD00100032
3-trifluoromethyl pyridine, pyridine, 3-trifluoromethyl, 3-trifluoromethyl-pyridine, m-trifluoromethylpyridine, alpha,alpha,alpha-trifluoro-3-picoline, pubchem8025, 3-cf3-pyridine, 5-trifluoromethylpyridine
3-(trifluoromethyl)pyridine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions