accessibility menu, dialog, popup

UserName

CAS RN 3796-23-4

IUPAC Name: 3-(trifluoromethyl)pyridine
Synonyms: 3-trifluoromethylpyridine
Molecular Weight (g/mol): 147.1
Molecular Formula: C6H4F3N
InChi Key: JTZSFNHHVULOGJ-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC=CN=C1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)