missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethyl)pyridine 98.0+%, TCI America™

Catalog No. T27865G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T27865G 5 g
T278625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T27865G Supplier TCI America Supplier No. T27865G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3796-23-4
Molecular Formula C6H4F3N
Molecular Weight (g/mol) 147.1
MDL Number MFCD00100032
InChI Key JTZSFNHHVULOGJ-UHFFFAOYSA-N
Synonym 3-trifluoromethyl pyridine, pyridine, 3-trifluoromethyl, 3-trifluoromethyl-pyridine, m-trifluoromethylpyridine, alpha,alpha,alpha-trifluoro-3-picoline, pubchem8025, 3-cf3-pyridine, 5-trifluoromethylpyridine
PubChem CID 77417
IUPAC Name 3-(trifluoromethyl)pyridine
SMILES C1=CC(=CN=C1)C(F)(F)F

Specifications

Color Yellow
Boiling Point 113°C
Quantity 5 g
UN Number 1993
Formula Weight 147.10
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-(Trifluoromethyl)pyridine