missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethyl)phenylacetonitrile 98.0+%, TCI America™

Catalog No. p-7137181
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T23675G 5 g
T236725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T23675G Supplier TCI America Supplier No. T23675G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2338-76-3
Molecular Formula C9H6F3N
Molecular Weight (g/mol) 185.15
MDL Number MFCD00001913
InChI Key JOIYKSLWXLFGGR-UHFFFAOYSA-N
Synonym 3-trifluoromethyl phenylacetonitrile, 3-trifluoromethylbenzylcyanide, 2-3-trifluoromethyl phenyl acetonitrile, benzeneacetonitrile, 3-trifluoromethyl, 3-trifluoromethyl benzyl cyanide, m-trifluoromethylphenylacetonitrile, m-trifluoromethylbenzyl cyanide, 3-trifluoromethylphenylacetonitrile, 3-trifluormethylbenzylcyanide, 3-trifluoromethyl benzeneacetonitrile
PubChem CID 75360
IUPAC Name 2-[3-(trifluoromethyl)phenyl]acetonitrile
SMILES FC(F)(F)C1=CC=CC(CC#N)=C1

Specifications

Color Yellow
Boiling Point 93°C
Quantity 5 g
UN Number 1992
Formula Weight 185.15
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-(Trifluoromethyl)phenylacetonitrile
RTECSNumber CY1820000
TSCA Yes