missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethyl)phenylacetic Acid 98.0+%, TCI America™

$70.84 - $283.33

Chemical Identifiers

CAS 351-35-9
Molecular Formula C9H7F3O2
Molecular Weight (g/mol) 204.148
MDL Number MFCD00004339
InChI Key BLXXCCIBGGBDHI-UHFFFAOYSA-N
Synonym 3-trifluoromethyl phenylacetic acid, m-trifluoromethyl phenylacetic acid, 2-3-trifluoromethyl phenyl acetic acid, benzeneacetic acid, 3-trifluoromethyl, 3-trifluoromethylphenylacetic acid, 3-trifluoromethyl phenyl acetic acid, a,a,a-trifluoro-m-tolyl acetic acid, alpha,alpha,alpha-trifluoro-m-tolyl acetic acid, 3-carboxymethyl benzotrifluoride
PubChem CID 67695
IUPAC Name 2-[3-(trifluoromethyl)phenyl]acetic acid
SMILES C1=CC(=CC(=C1)C(F)(F)F)CC(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
T16625G
View Documents
TCI America
T16625G
5 g N/A
Only null left
 
T166225G
View Documents
TCI America
T166225G
25 g N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

351-35-9
204.148
BLXXCCIBGGBDHI-UHFFFAOYSA-N
67695
C1=CC(=CC(=C1)C(F)(F)F)CC(=O)O
C9H7F3O2
MFCD00004339
3-trifluoromethyl phenylacetic acid, m-trifluoromethyl phenylacetic acid, 2-3-trifluoromethyl phenyl acetic acid, benzeneacetic acid, 3-trifluoromethyl, 3-trifluoromethylphenylacetic acid, 3-trifluoromethyl phenyl acetic acid, a,a,a-trifluoro-m-tolyl acetic acid, alpha,alpha,alpha-trifluoro-m-tolyl acetic acid, 3-carboxymethyl benzotrifluoride
2-[3-(trifluoromethyl)phenyl]acetic acid

Specifications

351-35-9
White-Yellow
MFCD00004339
3-trifluoromethyl phenylacetic acid, m-trifluoromethyl phenylacetic acid, 2-3-trifluoromethyl phenyl acetic acid, benzeneacetic acid, 3-trifluoromethyl, 3-trifluoromethylphenylacetic acid, 3-trifluoromethyl phenyl acetic acid, a,a,a-trifluoro-m-tolyl acetic acid, alpha,alpha,alpha-trifluoro-m-tolyl acetic acid, 3-carboxymethyl benzotrifluoride
C1=CC(=CC(=C1)C(F)(F)F)CC(=O)O
204.148
204.15
Crystalline Powder
77°C
C9H7F3O2
5 g
BLXXCCIBGGBDHI-UHFFFAOYSA-N
2-[3-(trifluoromethyl)phenyl]acetic acid
67695
≥98.0% (T)
3-(Trifluoromethyl)phenylacetic Acid
Videos
Safety and Handling

Safety and Handling

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'