Learn More
3-(Trifluoromethyl)phenylacetic acid, 96%
CAS: 351-35-9 | C9H7F3O2 | 204.148 g/mol
$165.77 - $591.38
Chemical Identifiers
| CAS | 351-35-9 |
|---|---|
| Molecular Formula | C9H7F3O2 |
| Molecular Weight (g/mol) | 204.148 |
| MDL Number | MFCD00004339 |
| InChI Key | BLXXCCIBGGBDHI-UHFFFAOYSA-N |
| Synonym | 3-trifluoromethyl phenylacetic acid, m-trifluoromethyl phenylacetic acid, 2-3-trifluoromethyl phenyl acetic acid, benzeneacetic acid, 3-trifluoromethyl, 3-trifluoromethylphenylacetic acid, 3-trifluoromethyl phenyl acetic acid, a,a,a-trifluoro-m-tolyl acetic acid, alpha,alpha,alpha-trifluoro-m-tolyl acetic acid, 3-carboxymethyl benzotrifluoride |
| PubChem CID | 67695 |
| IUPAC Name | 2-[3-(trifluoromethyl)phenyl]acetic acid |
| SMILES | C1=CC(=CC(=C1)C(F)(F)F)CC(=O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL1096009
|
Thermo Scientific Chemicals
L1096009 |
10 g |
Each for $165.77
|
|
|||||
|
AAL1096018
|
Thermo Scientific Chemicals
L1096018 |
50 g |
Each for $591.38
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 351-35-9 | |
| 204.148 | |
| BLXXCCIBGGBDHI-UHFFFAOYSA-N | |
| 67695 | |
| C1=CC(=CC(=C1)C(F)(F)F)CC(=O)O |
| C9H7F3O2 | |
| MFCD00004339 | |
| 3-trifluoromethyl phenylacetic acid, m-trifluoromethyl phenylacetic acid, 2-3-trifluoromethyl phenyl acetic acid, benzeneacetic acid, 3-trifluoromethyl, 3-trifluoromethylphenylacetic acid, 3-trifluoromethyl phenyl acetic acid, a,a,a-trifluoro-m-tolyl acetic acid, alpha,alpha,alpha-trifluoro-m-tolyl acetic acid, 3-carboxymethyl benzotrifluoride | |
| 2-[3-(trifluoromethyl)phenyl]acetic acid |
Specifications
| 351-35-9 | |
| C9H7F3O2 | |
| 10 g | |
| 3-trifluoromethyl phenylacetic acid, m-trifluoromethyl phenylacetic acid, 2-3-trifluoromethyl phenyl acetic acid, benzeneacetic acid, 3-trifluoromethyl, 3-trifluoromethylphenylacetic acid, 3-trifluoromethyl phenyl acetic acid, a,a,a-trifluoro-m-tolyl acetic acid, alpha,alpha,alpha-trifluoro-m-tolyl acetic acid, 3-carboxymethyl benzotrifluoride | |
| C1=CC(=CC(=C1)C(F)(F)F)CC(=O)O | |
| 204.148 | |
| 204.15 | |
| 3-(Trifluoromethyl)phenylacetic acid |
| 77°C to 78°C | |
| MFCD00004339 | |
| 2213223 | |
| BLXXCCIBGGBDHI-UHFFFAOYSA-N | |
| 2-[3-(trifluoromethyl)phenyl]acetic acid | |
| 67695 | |
| ≥98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 206-511-7
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only