missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethyl)phenyl Isocyanate 96.0+%, TCI America™

Catalog No. I03405G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I03405G 5 g
I034025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I03405G Supplier TCI America Supplier No. I03405G

Chemical Identifiers

CAS 329-01-1
Molecular Formula C8H4F3NO
Molecular Weight (g/mol) 187.12
MDL Number MFCD00002020
InChI Key SXJYSIBLFGQAND-UHFFFAOYSA-N
Synonym 3-trifluoromethyl phenyl isocyanate, 1-isocyanato-3-trifluoromethyl benzene, benzene, 1-isocyanato-3-trifluoromethyl, a,a,a-trifluoro-m-tolyl isocyanate, isocyanobenzotrifluoride, unii-qps7llz6xm, 3-trifluoromethylphenyl isocyanate, alpha,alpha,alpha-trifluoro-m-tolyl isocyanate, 3-trifluoromethyl phenylisocyanate, qps7llz6xm
PubChem CID 9483
IUPAC Name 1-isocyanato-3-(trifluoromethyl)benzene
SMILES FC(F)(F)C1=CC=CC(=C1)N=C=O

Specifications

Color Yellow
Boiling Point 104°C
Quantity 5 g
UN Number 2285
Formula Weight 187.12
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-(Trifluoromethyl)phenyl Isocyanate
RTECSNumber NR0200000
TSCA Yes
Recommended Storage Refrigerator