missing translation for 'onlineSavingsMsg'
Learn More
Learn More
[3-(Trifluoromethyl)phenyl](2,4,6-trimethylphenyl)iodonium Trifluoromethanesulfonate 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 183°C |
| Quantity | 5 g |
| Formula Weight | 540.26 |
| Percent Purity | ≥98.0% (HPLC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | [3-(Trifluoromethyl)phenyl](2,4,6-trimethylphenyl)iodonium Trifluoromethanesulfonate |
Safety and Handling
| TSCA | No |