missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethyl)phenol, 98+%

Catalog No. AAA1278406
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1278406 5 g
AAA1278414 25 g
AAA1278422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1278406 Supplier Thermo Scientific Chemicals Supplier No. A1278406
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 98-17-9 | C7H5F3O | 162.111 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 98-17-9
Molecular Formula C7H5F3O
Molecular Weight (g/mol) 162.111
MDL Number MFCD00002299
InChI Key UGEJOEBBMPOJMT-UHFFFAOYSA-N
Synonym 3-trifluoromethyl phenol, 3-hydroxybenzotrifluoride, m-trifluoromethylphenol, m-hydroxybenzotrifluoride, phenol, 3-trifluoromethyl, m-trifluoromethyl phenol, alpha,alpha,alpha-trifluoro-m-cresol, a,a,a-trifluoro-m-cresol
PubChem CID 7376
IUPAC Name 3-(trifluoromethyl)phenol
SMILES C1=CC(=CC(=C1)O)C(F)(F)F

Specifications

Melting Point -2°C
Density 1.336
Boiling Point 177°C to 178°C
Flash Point 74°C (165°F)
Odor Phenol-like
Refractive Index 1.458
Quantity 5 g
UN Number UN3265
Beilstein 2045663
Formula Weight 162.11
Percent Purity ≥98%
Chemical Name or Material 3-(Trifluoromethyl)phenol
Show More Show Less
Hazard Category H227-H302-H314-H335
Hazard Statement GHS H Statement
H314-H318-H302-H312-H332-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Combustible liquid.
Precautionary Statement P210-P235-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 202-645-5
RTECSNumber GP3510000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only