missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethyl)crotonic acid, 96%

Catalog No. p-7043976
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0381614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0381614 Supplier Thermo Scientific Chemicals Supplier No. L0381614

CAS: 69056-67-3 | C5H5F3O2 | 154.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 69056-67-3
Molecular Formula C5H5F3O2
Molecular Weight (g/mol) 154.09
MDL Number MFCD00014374,MFCD01457157
InChI Key QRRCTLYMABZQCS-NSCUHMNNSA-N
Synonym beta-trifluoromethylcrotonic acid, 3-trifluoromethylcrotonic acid, 3-trifluoromethyl crotonic acid, 2z-4,4,4-trifluoro-3-methylbut-2-enoic acid, heddpadiaicichiceiumlbbp, z-4,4,4-trifluoro-3-methylbut-2-enoic acid, 4,4,4-trifluoro-3-methyl-2-butenoic acid, 2-butenoic acid,4,4,4-trifluoro-3-methyl-, 2z
PubChem CID 5708835
IUPAC Name (Z)-4,4,4-trifluoro-3-methylbut-2-enoic acid
SMILES C\C(=C/C(O)=O)C(F)(F)F

Specifications

Melting Point 28°C to 30°C
Density 1.318
Boiling Point 158°C to 163°C
Flash Point 79°C (174°F)
Refractive Index 1.385
Quantity 25 g
UN Number UN3261
Beilstein 1769204
Formula Weight 154.09
Percent Purity 96%
Chemical Name or Material 3-(Trifluoromethyl)crotonic acid
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only