CAS RN 69056-67-3
IUPAC Name:
(2E)-4,4,4-trifluoro-3-methylbut-2-enoic acid
Synonyms:
(2E)-4,4,4-trifluoro-3-methylbut-2-enoic acid
Molecular Weight (g/mol):
154.09
Molecular Formula:
C5H5F3O2
InChi Key:
QRRCTLYMABZQCS-NSCUHMNNSA-N
SMILES:
C\C(=C/C(O)=O)C(F)(F)F
1
–
1
of
1
results
1
Promotions Available
3-(Trifluoromethyl)crotonic acid, 96%
3-(Trifluoromethyl)crotonic acid, 96%
CAS: 69056-67-3 Molecular Formula: C5H5F3O2 Molecular Weight (g/mol): 154.09 MDL Number: MFCD00014374,MFCD01457157 InChI Key: QRRCTLYMABZQCS-NSCUHMNNSA-N Synonym: beta-trifluoromethylcrotonic acid,3-trifluoromethylcrotonic acid,3-trifluoromethyl crotonic acid,2z-4,4,4-trifluoro-3-methylbut-2-enoic acid,heddpadiaicichiceiumlbbp,z-4,4,4-trifluoro-3-methylbut-2-enoic acid,4,4,4-trifluoro-3-methyl-2-butenoic acid,2-butenoic acid,4,4,4-trifluoro-3-methyl-, 2z PubChem CID: 5708835 IUPAC Name: (Z)-4,4,4-trifluoro-3-methylbut-2-enoic acid SMILES: C\C(=C/C(O)=O)C(F)(F)F