Learn More
3-(Trifluoromethyl)benzylamine, 98%, Thermo Scientific™
$153.54 - $663.81
Chemical Identifiers
| CAS | 2740-83-2 |
|---|---|
| Molecular Formula | C8H8F3N |
| Molecular Weight (g/mol) | 175.15 |
| MDL Number | MFCD00008117 |
| InChI Key | YKNZTUQUXUXTLE-UHFFFAOYSA-N |
| Synonym | 3-trifluoromethyl benzylamine, 3-trifluoromethyl phenyl methanamine, 3-trifluoromethylbenzylamine, m-trifluoromethylbenzylamine, m-trifluoromethylbenzyl amine, 3-trifluoromethyl benzyl amine, benzenemethanamine, 3-trifluoromethyl, 3-trifluoromethyl-benzylamine, m-trifluoromethyl benzylamine |
| PubChem CID | 75962 |
| IUPAC Name | [3-(trifluoromethyl)phenyl]methanamine |
| SMILES | C1=CC(=CC(=C1)C(F)(F)F)CN |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
AC312960050
|
N/A | 5g | Glass bottle |
Each for $153.54
|
|
||||
|
AC312960250
|
N/A | 25g | Glass bottle |
Each for $663.81
|
|
||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| C8H8F3N | |
| MFCD00008117 | |
| 3-trifluoromethyl benzylamine, 3-trifluoromethyl phenyl methanamine, 3-trifluoromethylbenzylamine, m-trifluoromethylbenzylamine, m-trifluoromethylbenzyl amine, 3-trifluoromethyl benzyl amine, benzenemethanamine, 3-trifluoromethyl, 3-trifluoromethyl-benzylamine, m-trifluoromethyl benzylamine | |
| [3-(trifluoromethyl)phenyl]methanamine |
Specifications
| 2740-83-2 | |
| 100.0 | |
| 93.0°C to 97.0°C (22.0mmHg) | |
| Authentic | |
| C8H8F3N | |
| CF3C6H4CH2NH2 | |
| 1.22 | |
| Solubility in water: insoluble | |
| C1=CC(=CC(=C1)C(F)(F)F)CN | |
| 175.15 | |
| 175.15 | |
| 3-(Trifluoromethyl)benzylamine |
| 97.5 | |
| 1.2200g/mL | |
| 71°C | |
| 97.5% min. (GC) | |
| 1.4620 to 1.4640 | |
| MFCD00008117 | |
| 3-trifluoromethyl benzylamine, 3-trifluoromethyl phenyl methanamine, 3-trifluoromethylbenzylamine, m-trifluoromethylbenzylamine, m-trifluoromethylbenzyl amine, 3-trifluoromethyl benzyl amine, benzenemethanamine, 3-trifluoromethyl, 3-trifluoromethyl-benzylamine, m-trifluoromethyl benzylamine | |
| YKNZTUQUXUXTLE-UHFFFAOYSA-N | |
| [3-(trifluoromethyl)phenyl]methanamine | |
| 75962 | |
| 98% |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
EINECSNumber : 220-367-2
RUO – Research Use Only