missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethyl)benzylamine, 98%

Catalog No. AC312960250
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC312960250 25 g Glass bottle
AC312960050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC312960250 Supplier Thermo Scientific Chemicals Supplier No. 312960250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2740-83-2 | C8H8F3N | 175.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2740-83-2
Molecular Formula C8H8F3N
Molecular Weight (g/mol) 175.15
MDL Number MFCD00008117
InChI Key YKNZTUQUXUXTLE-UHFFFAOYSA-N
Synonym 3-trifluoromethyl benzylamine, 3-trifluoromethyl phenyl methanamine, 3-trifluoromethylbenzylamine, m-trifluoromethylbenzylamine, m-trifluoromethylbenzyl amine, 3-trifluoromethyl benzyl amine, benzenemethanamine, 3-trifluoromethyl, 3-trifluoromethyl-benzylamine, m-trifluoromethyl benzylamine
PubChem CID 75962
IUPAC Name [3-(trifluoromethyl)phenyl]methanamine
SMILES C1=CC(=CC(=C1)C(F)(F)F)CN

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.2200g/mL
Boiling Point 93.0°C to 97.0°C (22.0 mmHg)
Flash Point 71°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4620 to 1.4640
Linear Formula CF3C6H4CH2NH2
Quantity 25 g
Specific Gravity 1.22
Solubility Information Solubility in water: insoluble
Formula Weight 175.15
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3-(Trifluoromethyl)benzylamine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
EINECSNumber 220-367-2

RUO – Research Use Only