Learn More
3-(Trifluoromethyl)benzylamine, 97%
CAS: 2740-83-2 | C8H8F3N | 175.154 g/mol
$112.62 - $559.60
Chemical Identifiers
| CAS | 2740-83-2 |
|---|---|
| Molecular Formula | C8H8F3N |
| Molecular Weight (g/mol) | 175.154 |
| MDL Number | MFCD00008117 |
| InChI Key | YKNZTUQUXUXTLE-UHFFFAOYSA-N |
| Synonym | 3-trifluoromethyl benzylamine, 3-trifluoromethyl phenyl methanamine, 3-trifluoromethylbenzylamine, m-trifluoromethylbenzylamine, m-trifluoromethylbenzyl amine, 3-trifluoromethyl benzyl amine, benzenemethanamine, 3-trifluoromethyl, 3-trifluoromethyl-benzylamine, m-trifluoromethyl benzylamine |
| PubChem CID | 75962 |
| IUPAC Name | [3-(trifluoromethyl)phenyl]methanamine |
| SMILES | C1=CC(=CC(=C1)C(F)(F)F)CN |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAB2414306
|
Thermo Scientific Chemicals
B2414306 |
5 g |
Each for $112.62
|
|
|||||
|
AAB2414314
|
Thermo Scientific Chemicals
B2414314 |
25 g |
Each for $559.60
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 2740-83-2 | |
| 175.154 | |
| YKNZTUQUXUXTLE-UHFFFAOYSA-N | |
| 75962 | |
| C1=CC(=CC(=C1)C(F)(F)F)CN |
| C8H8F3N | |
| MFCD00008117 | |
| 3-trifluoromethyl benzylamine, 3-trifluoromethyl phenyl methanamine, 3-trifluoromethylbenzylamine, m-trifluoromethylbenzylamine, m-trifluoromethylbenzyl amine, 3-trifluoromethyl benzyl amine, benzenemethanamine, 3-trifluoromethyl, 3-trifluoromethyl-benzylamine, m-trifluoromethyl benzylamine | |
| [3-(trifluoromethyl)phenyl]methanamine |
Specifications
| 2740-83-2 | |
| 93°C to 97°C (28 mmHg) | |
| C8H8F3N | |
| MFCD00008117 | |
| UN2735 | |
| Air Sensitive | |
| YKNZTUQUXUXTLE-UHFFFAOYSA-N | |
| [3-(trifluoromethyl)phenyl]methanamine | |
| 75962 | |
| 97% |
| 1.222 | |
| 71°C (159°F) | |
| 1.463 | |
| 5 g | |
| 1102465 | |
| 3-trifluoromethyl benzylamine, 3-trifluoromethyl phenyl methanamine, 3-trifluoromethylbenzylamine, m-trifluoromethylbenzylamine, m-trifluoromethylbenzyl amine, 3-trifluoromethyl benzyl amine, benzenemethanamine, 3-trifluoromethyl, 3-trifluoromethyl-benzylamine, m-trifluoromethyl benzylamine | |
| C1=CC(=CC(=C1)C(F)(F)F)CN | |
| 175.154 | |
| 175.15 | |
| 3-(Trifluoromethyl)benzylamine |
Safety and Handling
GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
P210-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H227-H314
DOTInformation : Hazard Class: 8; Packaging Group: II
EINECSNumber : 220-367-2
TSCA : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only