missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethyl)benzyl Chloride 95.0+%, TCI America™

Catalog No. T22905G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T22905G 5 g
T229025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T22905G Supplier TCI America Supplier No. T22905G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 705-29-3
Molecular Formula C8H6ClF3
Molecular Weight (g/mol) 194.581
InChI Key XGASTRVQNVVYIZ-UHFFFAOYSA-N
Synonym 3-trifluoromethyl benzyl chloride, 1-chloromethyl-3-trifluoromethyl benzene, 3-trifluoromethylbenzyl chloride, 3-chloromethyl benzotrifluoride, m-trifluoromethylbenzyl chloride, 3-chloromethyl-benzotrifluoride, benzene, 1-chloromethyl-3-trifluoromethyl, alpha'-chloro-alpha,alpha,alpha-trifluoro-m-xylene, 3-chloromethylbenzotrifluoride, 3-trifluoromethylbenzylchloride
PubChem CID 69716
IUPAC Name 1-(chloromethyl)-3-(trifluoromethyl)benzene
SMILES C1=CC(=CC(=C1)C(F)(F)F)CCl

Specifications

Color Colorless
Boiling Point 70°C
Quantity 5 g
UN Number 2920
Formula Weight 194.58
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-(Trifluoromethyl)benzyl Chloride