missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethyl)benzyl chloride, 98%

Catalog No. AAA1625322
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1625322 100 g
AAA1625306 5 g
AAA1625314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1625322 Supplier Thermo Scientific Chemicals Supplier No. A1625322
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 705-29-3 | C8H6ClF3 | 194.581 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 705-29-3
Molecular Formula C8H6ClF3
Molecular Weight (g/mol) 194.581
MDL Number MFCD00000908
InChI Key XGASTRVQNVVYIZ-UHFFFAOYSA-N
Synonym 3-trifluoromethyl benzyl chloride, 1-chloromethyl-3-trifluoromethyl benzene, 3-trifluoromethylbenzyl chloride, 3-chloromethyl benzotrifluoride, m-trifluoromethylbenzyl chloride, 3-chloromethyl-benzotrifluoride, benzene, 1-chloromethyl-3-trifluoromethyl, alpha'-chloro-alpha,alpha,alpha-trifluoro-m-xylene, 3-chloromethylbenzotrifluoride, 3-trifluoromethylbenzylchloride
PubChem CID 69716
IUPAC Name 1-(chloromethyl)-3-(trifluoromethyl)benzene
SMILES C1=CC(=CC(=C1)C(F)(F)F)CCl

Specifications

Density 1.254
Boiling Point 68°C to 70°C (12 mmHg)
Flash Point 74°C (165°F)
Odor Pungent
Refractive Index 1.4645
Quantity 100 g
UN Number UN3265
Beilstein 743242
Formula Weight 194.59
Percent Purity 98%
Chemical Name or Material 3-(Trifluoromethyl)benzyl chloride
Show More Show Less
Hazard Category H226-H314
Hazard Statement GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P233-P240-P241-P242-P243-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 211-884-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only