missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(Trifluoromethyl)benzyl Alcohol 98.0+%, TCI America™
$124.89 - $210.63
Chemical Identifiers
| CAS | 349-75-7 |
|---|---|
| Molecular Formula | C8H7F3O |
| Molecular Weight (g/mol) | 176.138 |
| MDL Number | MFCD00004645 |
| InChI Key | BXEHKCUWIODEDE-UHFFFAOYSA-N |
| Synonym | 3-trifluoromethyl benzyl alcohol, 3-trifluoromethylbenzyl alcohol, 3-trifluoromethyl phenyl methanol, 3-trifluoromethyl benzylalcohol, m-trifluoromethylbenzyl alcohol, m-trifluoromethyl benzyl alcohol, 3-trifluoromethyl benzylic alcohol, benzenemethanol, 3-trifluoromethyl, m-trifluorome benzyl alcohol |
| PubChem CID | 67681 |
| IUPAC Name | [3-(trifluoromethyl)phenyl]methanol |
| SMILES | C1=CC(=CC(=C1)C(F)(F)F)CO |
Chemical Identifiers
| 349-75-7 | |
| 176.138 | |
| BXEHKCUWIODEDE-UHFFFAOYSA-N | |
| 67681 | |
| C1=CC(=CC(=C1)C(F)(F)F)CO |
| C8H7F3O | |
| MFCD00004645 | |
| 3-trifluoromethyl benzyl alcohol, 3-trifluoromethylbenzyl alcohol, 3-trifluoromethyl phenyl methanol, 3-trifluoromethyl benzylalcohol, m-trifluoromethylbenzyl alcohol, m-trifluoromethyl benzyl alcohol, 3-trifluoromethyl benzylic alcohol, benzenemethanol, 3-trifluoromethyl, m-trifluorome benzyl alcohol | |
| [3-(trifluoromethyl)phenyl]methanol |
Specifications
| 349-75-7 | |
| C8H7F3O | |
| 10 g | |
| BXEHKCUWIODEDE-UHFFFAOYSA-N | |
| [3-(trifluoromethyl)phenyl]methanol | |
| 67681 | |
| ≥98.0% (GC) | |
| 3-(Trifluoromethyl)benzyl Alcohol |
| 68°C | |
| MFCD00004645 | |
| 3-trifluoromethyl benzyl alcohol, 3-trifluoromethylbenzyl alcohol, 3-trifluoromethyl phenyl methanol, 3-trifluoromethyl benzylalcohol, m-trifluoromethylbenzyl alcohol, m-trifluoromethyl benzyl alcohol, 3-trifluoromethyl benzylic alcohol, benzenemethanol, 3-trifluoromethyl, m-trifluorome benzyl alcohol | |
| C1=CC(=CC(=C1)C(F)(F)F)CO | |
| 176.138 | |
| 176.14 | |
| Liquid |
Safety and Handling
TSCA : No