missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethyl)benzyl Alcohol 98.0+%, TCI America™

Catalog No. T134210G
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T134210G 10 g
T134225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T134210G Supplier TCI America Supplier No. T134210G

Chemical Identifiers

CAS 349-75-7
Molecular Formula C8H7F3O
Molecular Weight (g/mol) 176.138
MDL Number MFCD00004645
InChI Key BXEHKCUWIODEDE-UHFFFAOYSA-N
Synonym 3-trifluoromethyl benzyl alcohol, 3-trifluoromethylbenzyl alcohol, 3-trifluoromethyl phenyl methanol, 3-trifluoromethyl benzylalcohol, m-trifluoromethylbenzyl alcohol, m-trifluoromethyl benzyl alcohol, 3-trifluoromethyl benzylic alcohol, benzenemethanol, 3-trifluoromethyl, m-trifluorome benzyl alcohol
PubChem CID 67681
IUPAC Name [3-(trifluoromethyl)phenyl]methanol
SMILES C1=CC(=CC(=C1)C(F)(F)F)CO

Specifications

Boiling Point 68°C
Quantity 10 g
Formula Weight 176.14
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-(Trifluoromethyl)benzyl Alcohol