Learn More
3-(Trifluoromethyl)benzyl alcohol, 97%
CAS: 349-75-7 | C8H7F3O | 176.138 g/mol
$92.15 - $278.92
Chemical Identifiers
| CAS | 349-75-7 |
|---|---|
| Molecular Formula | C8H7F3O |
| Molecular Weight (g/mol) | 176.138 |
| MDL Number | MFCD00004645 |
| InChI Key | BXEHKCUWIODEDE-UHFFFAOYSA-N |
| Synonym | 3-trifluoromethyl benzyl alcohol, 3-trifluoromethylbenzyl alcohol, 3-trifluoromethyl phenyl methanol, 3-trifluoromethyl benzylalcohol, m-trifluoromethylbenzyl alcohol, m-trifluoromethyl benzyl alcohol, 3-trifluoromethyl benzylic alcohol, benzenemethanol, 3-trifluoromethyl, m-trifluorome benzyl alcohol |
| PubChem CID | 67681 |
| IUPAC Name | [3-(trifluoromethyl)phenyl]methanol |
| SMILES | C1=CC(=CC(=C1)C(F)(F)F)CO |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1673906
|
Thermo Scientific Chemicals
A1673906 |
5 g |
N/A
|
|
|||||
|
AAA1673914
|
Thermo Scientific Chemicals
A1673914 |
25 g |
N/A
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 349-75-7 | |
| 176.138 | |
| BXEHKCUWIODEDE-UHFFFAOYSA-N | |
| 67681 | |
| C1=CC(=CC(=C1)C(F)(F)F)CO |
| C8H7F3O | |
| MFCD00004645 | |
| 3-trifluoromethyl benzyl alcohol, 3-trifluoromethylbenzyl alcohol, 3-trifluoromethyl phenyl methanol, 3-trifluoromethyl benzylalcohol, m-trifluoromethylbenzyl alcohol, m-trifluoromethyl benzyl alcohol, 3-trifluoromethyl benzylic alcohol, benzenemethanol, 3-trifluoromethyl, m-trifluorome benzyl alcohol | |
| [3-(trifluoromethyl)phenyl]methanol |
Specifications
| 349-75-7 | |
| 79°C to 80°C (3 mmHg) | |
| C8H7F3O | |
| MFCD00004645 | |
| 1450161 | |
| BXEHKCUWIODEDE-UHFFFAOYSA-N | |
| [3-(trifluoromethyl)phenyl]methanol | |
| 67681 | |
| 97% |
| 1.293 | |
| 84°C (183°F) | |
| 1.46 | |
| 5 g | |
| 3-trifluoromethyl benzyl alcohol, 3-trifluoromethylbenzyl alcohol, 3-trifluoromethyl phenyl methanol, 3-trifluoromethyl benzylalcohol, m-trifluoromethylbenzyl alcohol, m-trifluoromethyl benzyl alcohol, 3-trifluoromethyl benzylic alcohol, benzenemethanol, 3-trifluoromethyl, m-trifluorome benzyl alcohol | |
| C1=CC(=CC(=C1)C(F)(F)F)CO | |
| 176.138 | |
| 176.14 | |
| 3-(Trifluoromethyl)benzyl alcohol |
Safety and Handling
GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H227-H315-H319-H335
EINECSNumber : 206-489-9
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only