missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethyl)benzyl alcohol, 97%

Catalog No. AAA1673906
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1673906 5 g
AAA1673914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1673906 Supplier Thermo Scientific Chemicals Supplier No. A1673906
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 349-75-7 | C8H7F3O | 176.138 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 349-75-7
Molecular Formula C8H7F3O
Molecular Weight (g/mol) 176.138
MDL Number MFCD00004645
InChI Key BXEHKCUWIODEDE-UHFFFAOYSA-N
Synonym 3-trifluoromethyl benzyl alcohol, 3-trifluoromethylbenzyl alcohol, 3-trifluoromethyl phenyl methanol, 3-trifluoromethyl benzylalcohol, m-trifluoromethylbenzyl alcohol, m-trifluoromethyl benzyl alcohol, 3-trifluoromethyl benzylic alcohol, benzenemethanol, 3-trifluoromethyl, m-trifluorome benzyl alcohol
PubChem CID 67681
IUPAC Name [3-(trifluoromethyl)phenyl]methanol
SMILES C1=CC(=CC(=C1)C(F)(F)F)CO

Specifications

Density 1.293
Boiling Point 79°C to 80°C (3 mmHg)
Flash Point 84°C (183°F)
Refractive Index 1.46
Quantity 5 g
Beilstein 1450161
Formula Weight 176.14
Percent Purity 97%
Chemical Name or Material 3-(Trifluoromethyl)benzyl alcohol
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
EINECSNumber 206-489-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only