missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethyl)benzoyl chloride, 97%

Catalog No. AAA1449322
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1449322 100 g
AAA1449306 5 g
AAA1449314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1449322 Supplier Thermo Scientific Chemicals Supplier No. A1449322
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2251-65-2 | C8H4ClF3O | 208.564 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2251-65-2
Molecular Formula C8H4ClF3O
Molecular Weight (g/mol) 208.564
MDL Number MFCD00000680
InChI Key RUJYJCANMOTJMO-UHFFFAOYSA-N
Synonym 3-trifluoromethyl benzoyl chloride, benzoyl chloride, 3-trifluoromethyl, m-trifluoromethyl benzoyl chloride, m-trifluoromethylbenzoyl chloride, alpha,alpha,alpha-trifluoro-m-toluoyl chloride, 3-trifluoromethyl benzoylchloride, 3-trifluoromethyl-benzoyl chloride, alpha,alpha,alpha-trifluoro-m-tolyl chloride, pubchem5468
PubChem CID 75257
IUPAC Name 3-(trifluoromethyl)benzoyl chloride
SMILES C1=CC(=CC(=C1)C(F)(F)F)C(=O)Cl

Specifications

Density 1.408
Boiling Point 189°C to 191°C
Flash Point >110°C (230°F)
Odor Pungent
Refractive Index 1.477
Quantity 100 g
UN Number UN3265
Beilstein 391266
Sensitivity Moisture sensitive
Formula Weight 208.57
Percent Purity 97%
Chemical Name or Material 3-(Trifluoromethyl)benzoyl chloride
Show More Show Less
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 218-844-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only