missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(Trifluoromethyl)benzoic Acid 98.0+%, TCI America™
$54.06 - $186.41
Chemical Identifiers
| CAS | 454-92-2 |
|---|---|
| Molecular Formula | C8H5F3O2 |
| Molecular Weight (g/mol) | 190.12 |
| MDL Number | MFCD00002519 |
| InChI Key | FQXQBFUUVCDIRK-UHFFFAOYSA-N |
| Synonym | 3-trifluoromethyl benzoic acid, m-trifluoromethylbenzoic acid, alpha,alpha,alpha-trifluoro-m-toluic acid, 3-carboxybenzotrifluoride, 3-trifluoromethyl-benzoic acid, benzoic acid, 3-trifluoromethyl, m-trifluoromethyl benzoic acid, 3-trifluoromethyl benzoicacid, m-carboxybenzotrifluoride, a,a,a-trifluoro-m-toluic acid |
| PubChem CID | 9963 |
| ChEBI | CHEBI:60695 |
| IUPAC Name | 3-(trifluoromethyl)benzoic acid |
| SMILES | OC(=O)C1=CC=CC(=C1)C(F)(F)F |
Chemical Identifiers
| 454-92-2 | |
| 190.12 | |
| FQXQBFUUVCDIRK-UHFFFAOYSA-N | |
| 9963 | |
| 3-(trifluoromethyl)benzoic acid |
| C8H5F3O2 | |
| MFCD00002519 | |
| 3-trifluoromethyl benzoic acid, m-trifluoromethylbenzoic acid, alpha,alpha,alpha-trifluoro-m-toluic acid, 3-carboxybenzotrifluoride, 3-trifluoromethyl-benzoic acid, benzoic acid, 3-trifluoromethyl, m-trifluoromethyl benzoic acid, 3-trifluoromethyl benzoicacid, m-carboxybenzotrifluoride, a,a,a-trifluoro-m-toluic acid | |
| CHEBI:60695 | |
| OC(=O)C1=CC=CC(=C1)C(F)(F)F |
Specifications
| 454-92-2 | |
| White | |
| C8H5F3O2 | |
| 5 g | |
| FQXQBFUUVCDIRK-UHFFFAOYSA-N | |
| 3-(trifluoromethyl)benzoic acid | |
| 9963 | |
| 190.12 | |
| Crystalline Powder |
| 105°C | |
| 126°C | |
| MFCD00002519 | |
| 3-trifluoromethyl benzoic acid, m-trifluoromethylbenzoic acid, alpha,alpha,alpha-trifluoro-m-toluic acid, 3-carboxybenzotrifluoride, 3-trifluoromethyl-benzoic acid, benzoic acid, 3-trifluoromethyl, m-trifluoromethyl benzoic acid, 3-trifluoromethyl benzoicacid, m-carboxybenzotrifluoride, a,a,a-trifluoro-m-toluic acid | |
| OC(=O)C1=CC=CC(=C1)C(F)(F)F | |
| 190.12 | |
| CHEBI:60695 | |
| ≥98.0% (GC,T) | |
| 3-(Trifluoromethyl)benzoic Acid |
Safety and Handling
TSCA : No