missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethyl)benzoic acid, 98+%

Catalog No. AAA1673709
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1673709 10 g
AAA1673718 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1673709 Supplier Thermo Scientific Chemicals Supplier No. A1673709
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 454-92-2 | C8H5F3O2 | 190.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 454-92-2
Molecular Formula C8H5F3O2
Molecular Weight (g/mol) 190.12
MDL Number MFCD00002519
InChI Key FQXQBFUUVCDIRK-UHFFFAOYSA-N
Synonym 3-trifluoromethyl benzoic acid, m-trifluoromethylbenzoic acid, alpha,alpha,alpha-trifluoro-m-toluic acid, 3-carboxybenzotrifluoride, 3-trifluoromethyl-benzoic acid, benzoic acid, 3-trifluoromethyl, m-trifluoromethyl benzoic acid, 3-trifluoromethyl benzoicacid, m-carboxybenzotrifluoride, a,a,a-trifluoro-m-toluic acid
PubChem CID 9963
ChEBI CHEBI:60695
IUPAC Name 3-(trifluoromethyl)benzoic acid
SMILES OC(=O)C1=CC=CC(=C1)C(F)(F)F

Specifications

Melting Point 103°C to 107°C
Boiling Point 237°C to 240°C
Quantity 10 g
Beilstein 2049239
Formula Weight 190.12
Percent Purity ≥98%
Chemical Name or Material 3-(Trifluoromethyl)benzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 207-230-2
TSCA Yes

RUO – Research Use Only