missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethyl)benzeneboronic acid, 98%

CAS: 1423-26-3 | C7H6BF3O2 | 189.93 g/mol

Supplier:  Thermo Scientific Chemicals B2166106

 View more versions of this product

Catalog No. AAB2166106


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3-(Trifluoromethyl)benzeneboronic acid
162°C to 166°C
MFCD00151854
6084746
WOAORAPRPVIATR-UHFFFAOYSA-N
[3-(trifluoromethyl)phenyl]boronic acid
2734388
98%
1423-26-3
C7H6BF3O2
5 g
3-trifluoromethyl phenylboronic acid, 3-trifluoromethylphenylboronic acid, 3-trifluoromethyl phenyl boronic acid, 3-trifluoromethyl benzeneboronic acid, m-trifluoromethyl phenylboronic acid, 3-trifluoromethylboronic acid, 3-trifluoromethyl phenyl boranediol, 3-boronobenzotrifluoride, 3-trifluoromethyl benzeneboronic acid/anhydride
OB(O)C1=CC=CC(=C1)C(F)(F)F
189.93
189.93

Chemical Identifiers

1423-26-3
189.93
WOAORAPRPVIATR-UHFFFAOYSA-N
2734388
OB(O)C1=CC=CC(=C1)C(F)(F)F
C7H6BF3O2
MFCD00151854
3-trifluoromethyl phenylboronic acid, 3-trifluoromethylphenylboronic acid, 3-trifluoromethyl phenyl boronic acid, 3-trifluoromethyl benzeneboronic acid, m-trifluoromethyl phenylboronic acid, 3-trifluoromethylboronic acid, 3-trifluoromethyl phenyl boranediol, 3-boronobenzotrifluoride, 3-trifluoromethyl benzeneboronic acid/anhydride
[3-(trifluoromethyl)phenyl]boronic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only