missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethyl)benzeneboronic acid, 98%

Catalog No. AAB2166103
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2166103 1 g
AAB2166106 5 g
AAB2166114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2166103 Supplier Thermo Scientific Chemicals Supplier No. B2166103
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1423-26-3 | C7H6BF3O2 | 189.93 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1423-26-3
Molecular Formula C7H6BF3O2
Molecular Weight (g/mol) 189.93
MDL Number MFCD00151854
InChI Key WOAORAPRPVIATR-UHFFFAOYSA-N
Synonym 3-trifluoromethyl phenylboronic acid, 3-trifluoromethylphenylboronic acid, 3-trifluoromethyl phenyl boronic acid, 3-trifluoromethyl benzeneboronic acid, m-trifluoromethyl phenylboronic acid, 3-trifluoromethylboronic acid, 3-trifluoromethyl phenyl boranediol, 3-boronobenzotrifluoride, 3-trifluoromethyl benzeneboronic acid/anhydride
PubChem CID 2734388
IUPAC Name [3-(trifluoromethyl)phenyl]boronic acid
SMILES OB(O)C1=CC=CC(=C1)C(F)(F)F

Specifications

Melting Point 162°C to 166°C
Quantity 1 g
Beilstein 6084746
Formula Weight 189.93
Percent Purity 98%
Chemical Name or Material 3-(Trifluoromethyl)benzeneboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only