Learn More
3-(Trifluoromethyl)benzaldehyde, 97%
CAS: 454-89-7 | C8H5F3O | 174.122 g/mol
Supplier: Thermo Scientific Chemicals A1046314
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 3- (Trifluoromethyl)benzaldehyde | |
| 1.304 | |
| 68°C (154°F) | |
| C8H5F3O | |
| MFCD00003373 | |
| 2327537 | |
| 3-trifluoromethyl benzaldehyde, m-trifluoromethylbenzaldehyde, benzaldehyde, 3-trifluoromethyl, alpha,alpha,alpha-trifluoro-m-tolualdehyde, alpha,alpha,alpha-trifluoro-3-tolualdehyde, unii-at7h4h5u3e, a,a,a-trifluoro-m-tolualdehyde, 3-trifluoromethyl-benzaldehyde, mtf-bad, 3-formylbenzotrifluoride | |
| C1=CC(=CC(=C1)C(F)(F)F)C=O | |
| 174.122 | |
| 174.12 |
| 454-89-7 | |
| 83°C to 86°C (30 mmHg) | |
| Almond-like | |
| 1.465 | |
| 25 g | |
| Air Sensitive | |
| NMTUHPSKJJYGML-UHFFFAOYSA-N | |
| 3-(trifluoromethyl)benzaldehyde | |
| 67990 | |
| 97% |
Chemical Identifiers
| 454-89-7 | |
| 174.122 | |
| NMTUHPSKJJYGML-UHFFFAOYSA-N | |
| 67990 | |
| C1=CC(=CC(=C1)C(F)(F)F)C=O |
| C8H5F3O | |
| MFCD00003373 | |
| 3-trifluoromethyl benzaldehyde, m-trifluoromethylbenzaldehyde, benzaldehyde, 3-trifluoromethyl, alpha,alpha,alpha-trifluoro-m-tolualdehyde, alpha,alpha,alpha-trifluoro-3-tolualdehyde, unii-at7h4h5u3e, a,a,a-trifluoro-m-tolualdehyde, 3-trifluoromethyl-benzaldehyde, mtf-bad, 3-formylbenzotrifluoride | |
| 3-(trifluoromethyl)benzaldehyde |
Safety and Handling
GHS H Statement
H302-H315-H319-H335-H227
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H227-H315-H319-H335
EINECSNumber : 207-228-1
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only