missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethyl)-5,6,7,8-tetrahydro-1,2,4-triazolo[4,3-a]pyrazine hydrochloride, 98+%

Catalog No. AAH6499303
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6499303 1 g
AAH6499306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6499303 Supplier Thermo Scientific Chemicals Supplier No. H6499303
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 762240-92-6 | C6H8ClF3N4 | 228.603 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 762240-92-6
Molecular Formula C6H8ClF3N4
Molecular Weight (g/mol) 228.603
MDL Number MFCD09817638
InChI Key AQCSCRYRCRORET-UHFFFAOYSA-N
Synonym 3-trifluoromethyl-5,6,7,8-tetrahydro-1,2,4 triazolo 4,3-a pyrazine hydrochloride, 3-trifluoromethyl-5h,6h,7h,8h-1,2,4 triazolo 4,3-a pyrazine hydrochloride, 3-trifluoromethyl-5,6,7,8-tetrahydro-1,2,4-triazolo 4,3-a pyrazine hydrochloride, 3-trifluoromethyl-1,2,4-triazolo 4,3-a piperazine hydrochloride, 3-trifluoromethyl-5,6,7,8-tetrahydro-1,2,4 triazolo 4,3-a pyrazine hcl, 3-trifluoromethyl-5,6,7,8-tetrahydro-1,2,4-triazolo-4,3-a pyrazine hydrochloride, 3-trifluoromethyl-5,6,7,8-tetrahydro 1,2,4 triazolo 4,3-a pyrazine hydrochloride, 5,6,7,8-tetrahydro-3-trifluormethyl-4,3-a-1,2,4-triazolopyrazine hydrochloride, pubchem18203
PubChem CID 11961371
IUPAC Name 3-(trifluoromethyl)-5,6,7,8-tetrahydro-[1,2,4]triazolo[4,3-a]pyrazine;hydrochloride
SMILES C1CN2C(=NN=C2C(F)(F)F)CN1.Cl

Specifications

Quantity 1 g
Beilstein 9872881
Formula Weight 228.6
Percent Purity ≥98%
Chemical Name or Material 3-(Trifluoromethyl)-5,6,7,8-tetrahydro-1,2,4-triazolo[4,3-a]pyrazine hydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only