accessibility menu, dialog, popup

UserName

CAS RN 762240-92-6

IUPAC Name: 3-(trifluoromethyl)-5H,6H,7H,8H-[1,2,4]triazolo[4,3-a]pyrazine hydrochloride
Synonyms: 3-(trifluoromethyl)-5H,6H,7H,8H-[1,2,4]triazolo[4,3-a]pyrazine hydrochloride
Molecular Weight (g/mol): 228.6
Molecular Formula: C6H8ClF3N4
InChi Key: AQCSCRYRCRORET-UHFFFAOYSA-N
SMILES: Cl.FC(F)(F)C1=NN=C2CNCCN12
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)