missing translation for 'onlineSavingsMsg'
Learn More

3-Trifluoromethyl-4'-methoxydiphenylamine, TCI America™

Catalog No. T13261G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T13261G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T13261G Supplier TCI America Supplier No. T13261G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1494-26-4
Molecular Formula C14H12F3NO
Molecular Weight (g/mol) 267.25
MDL Number MFCD00143198
InChI Key VODIERGLAGAWQU-UHFFFAOYSA-N
Synonym N-(3-Trifluoromethylphenyl)-p-anisidine
PubChem CID 619960
IUPAC Name 4-methoxy-N-[3-(trifluoromethyl)phenyl]aniline
SMILES COC1=CC=C(NC2=CC=CC(=C2)C(F)(F)F)C=C1

Specifications

Melting Point 58°C
Color White
Quantity 1 g
Formula Weight 267.25
Physical Form Crystalline Powder
Chemical Name or Material 3-Trifluoromethyl-4′-methoxydiphenylamine