missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethyl)-1H-pyrazole, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 457340250

Quantity 25g

 View more versions of this product

Catalog No. AC457340250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

20154-03-4
136.08
3-trifluoromethyl pyrazole, 3-trifluoromethyl-1h-pyrazole, 5-trifluoromethyl-1h-pyrazole, 3-trifluoromethylpyrazole, trifluoromethylpyrazole, 1h-pyrazole, 3-trifluoromethyl, 3-trifluoromethyl-2h-pyrazole, pubchem10164, pubchem10423
5-(trifluoromethyl)-1H-pyrazole
C4H3F3N2
PYXNITNKYBLBMW-UHFFFAOYSA-N
1807034
C1=C(NN=C1)C(F)(F)F

Specifications

3-(Trifluoromethyl)-1H-pyrazole
70.0°C (0.5 mmHg)
96% min. (GC)
25g
PYXNITNKYBLBMW-UHFFFAOYSA-N
5-(trifluoromethyl)-1H-pyrazole
1807034
97%
20154-03-4
Authentic
C4H3F3N2
3-trifluoromethyl pyrazole, 3-trifluoromethyl-1h-pyrazole, 5-trifluoromethyl-1h-pyrazole, 3-trifluoromethylpyrazole, trifluoromethylpyrazole, 1h-pyrazole, 3-trifluoromethyl, 3-trifluoromethyl-2h-pyrazole, pubchem10164, pubchem10423
C1=C(NN=C1)C(F)(F)F
136.08
136.08
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a

Warning

SDS
Documents

Documents