missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethyl)-1H-pyrazole, 97%, Thermo Scientific™

Catalog No. p-7077668
Change view
Click to view available options
Quantity:
5g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC457340050 5g
AC457340250 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC457340050 Supplier Thermo Scientific Chemicals Supplier No. 457340050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 20154-03-4
Molecular Formula C4H3F3N2
Molecular Weight (g/mol) 136.08
InChI Key PYXNITNKYBLBMW-UHFFFAOYSA-N
Synonym 3-trifluoromethyl pyrazole, 3-trifluoromethyl-1h-pyrazole, 5-trifluoromethyl-1h-pyrazole, 3-trifluoromethylpyrazole, trifluoromethylpyrazole, 1h-pyrazole, 3-trifluoromethyl, 3-trifluoromethyl-2h-pyrazole, pubchem10164, pubchem10423
PubChem CID 1807034
IUPAC Name 5-(trifluoromethyl)-1H-pyrazole
SMILES C1=C(NN=C1)C(F)(F)F

Specifications

Boiling Point 70.0°C (0.5 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Quantity 5g
Formula Weight 136.08
Percent Purity 97%
Chemical Name or Material 3-(Trifluoromethyl)-1H-pyrazole
Hazard Category Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a

RUO – Research Use Only