missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethoxy)thiophenol, 98%

Catalog No. AAL1991203
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1991203 1 g
AAL1991206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1991203 Supplier Thermo Scientific Chemicals Supplier No. L1991203
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 220239-66-7 | C7H5F3OS | 194.171 g/mol

3-(Trifluoromethoxy)thiophenol is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-(Trifluoromethoxy)thiophenol is used as pharmaceutical intermediate.

Solubility
Not miscible in water.

Notes
Air sensitive. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 220239-66-7
Molecular Formula C7H5F3OS
Molecular Weight (g/mol) 194.171
MDL Number MFCD01320799
InChI Key GEJGGOYNWFQKKH-UHFFFAOYSA-N
PubChem CID 2777357
IUPAC Name 3-(trifluoromethoxy)benzenethiol
SMILES C1=CC(=CC(=C1)S)OC(F)(F)F

Specifications

Refractive Index 1.477
Quantity 1 g
UN Number UN2810
Sensitivity Air Sensitive
Synonym 3-trifluoromethoxy thiophenol, 3-trifluoromethoxy benzenethiol, 3-trifluoromethoxy benzene-1-thiol, benzenethiol, 3-trifluoromethoxy, acmc-1cejb, 3-trifluoromethoxythiophenol, 3-trifluoromethoxy benzene thiol, 3-trifluoromethyloxy benzenethiol, benzenethiol,3-trifluoromethoxy, 3-thio-alpha,alpha,alpha-trifluoroanisole
Solubility Information Not miscible in water.
Formula Weight 194.18
Percent Purity 98%
Chemical Name or Material 3-(Trifluoromethoxy)thiophenol
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only